Find methods for your needs
The application of SOLAμ for reduced sample volume analysis demonstrated with Niflumic acid. Be the first to rate this application
application solaμ for reduced sample volume analysis demonstrated with niflumic acid
# Compound Name Compound Class RT (min)
1 Niflumic acid Cyclooxygenase-2 Inhibitor 1.42
Description
The Thermo Scientific Dionex Ultimate 3000 system is applied to demonstrate how the Thermo Scientific SOLAμ SPE product can be used to facilitate the scale down of an extraction method when sample volume is limited. The Thermo Scientific Accucore HPLC column provided fast and efficient separation without the need for an ultra high pressure system. MS/MS detection was performed on a Thermo Scientific TSQ Vantage mass spectrometer. For Research Use Only. Not for use in diagnostic procedures.
Market: Pharma
Keywords: matrix effects, Micro-elution, niflumic acid, no dry down, reduced sample volume, reproducibility, SPE
Matrix: Human plasma
Uploaded on 10/21/2014.
For Research Use Only. Not for use in diagnostic procedures.