Find methods for your needs
AN225: Rapid Method for the Estimation of Total Free Monosaccharide Content of Corn Stover Hydrolysate Using HPAE-PAD
Description
Corn stover, an acid-hydrolyzed leaf, husk, stalk, and cob remenants in the field after harvest, is a common feedstock for fermentation systems used in biofuel production. HPAE-PAD is a well-accepted method for carbohydrate determinations such as those found corn stover. The CarboPac® PA1 anion-exchange column rapidly elutes corn stover carbohydrates, resolving the monosacccharides from the unretained and undetected noncarbohydrate components of acid-hydrolyzed corn stover. Under these conditions, disaccharides and trisaccharides are also resolved, with a total elution time less than 10 min.| Market: | Chemical |
| Keywords: | Arabinose, Cellobiose, corn stover, Fructose, galactose, glucose, Mannose, xylose |
| Matrix: | Corn stover (acid hydrolyzed) |
For Research Use Only. Not for use in diagnostic procedures.