Find methods for your needs
Determination of fenoprofen using accelerated conditions based on the USP method
Description
The Thermo Scientific HPLC system is applied for the determination of fenoprofen. The separation was performed on a Thermo Scientific Accucore RP-MS column, with a run time of 7 minutes and UV detection at 272 nm. The method of analysis is based on the USP monograph, and was scaled down by using an in-house method transfer calculator.| Market: | Pharma |
| Keywords: | Accucore RP-MS, Fenoprofen, Gemfibrozil, HPLC, NSAID, USP Monograph, UV detection |
| Matrix: | N/A |
| Author: | Monica Dolci, Derek Hillbeck |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.