Find methods for your needs
Improved analysis of four peptide maps on a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 RSLC system is applied for the analysis of peptide mapping. The separation of four peptide maps was achieved on a Thermo Scientific Acclaim 120 C18 column in 50 minutes with UV detection.| Market: | BioPharma |
| Keywords: | Acclaim 120 C18, beta-Casein, Bovine serum albumin, Fetuin, HPLC, Ovalbumin, Peptide maps, UV detection |
| Matrix: | Protein |
For Research Use Only. Not for use in diagnostic procedures.