Find methods for your needs
Comparison of a Thermo Scientific Acclaim Mixed-Mode HILIC-1 column against conventional RP and diol columns for characterization of an ethoxylated surfactant
Description
The Thermo Scientific Acclaim Mixed-Mode HILIC-1 column is compared against conventional reversed-phase and diol columns for the analysis of ethoxylated surfactants. The unique balance of hydrophilic and lipophilic functionality of the Acclaim Mixed-Mode HILIC-1 column provides excellent resolution of ethylene glycol oligomer distribution, unmatched by any conventional column.| Market: | Material Science |
| Keywords: | Acclaim Mixed-Mode HILIC-1, Comparison, Diol, EO, Ethoxylated surfactant, Ethylene glycol oligomers, HILIC, HPLC, Reversed-phase, UV |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.