Find methods for your needs
Improved separation of selected amino acids on a Thermo Scientific Acclaim Mixed-Mode WAX-1 column in HILIC mode
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 column is used for the separation of six selected amino acids, with UV detection at 210nm. The analysis is performed in HILIC mode, and the amino acids elute in order of most hydrophobic to least hydrophobic.| Market: | Pharma |
| Keywords: | Acclaim Mixed-Mode WAX-1, Alanine, Amino acids, Glycine, HILIC, HPLC, Isoleucine, Leucine, Serine, UV detection, Valine |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.