Find methods for your needs
Improved separation of active ingredients in a cough syrup using a Thermo Scientific Acclaim Mixed-Mode WAX-1 column
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 column is used for the separation of active ingredients in a cough syrup, with UV detection at 210 nm. The Thermo Scientific Acclaim Mixed-Mode WAX-1 column resolves the active ingredients under simple isocratic conditions.| Market: | Pharma |
| Keywords: | Acclaim Mixed-Mode WAX-1, Acetaminophen, Citrate, Cough syrup, Dextromethorphan, Doxylamine, HPLC, Isocratic, Pharmaceutical, Pseudoephedrine, UV detection |
| Matrix: | Cough syrup |
For Research Use Only. Not for use in diagnostic procedures.