Find methods for your needs
Ultrafast determination of chlorophyll on a Thermo Scientific Acclaim PolarAdvantage II (PA2) RSLC column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of chlorophyll. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage II (PA2) RSLC 2.2 µm column in under 2 minutes, with UV detection at 445 and 655 nm.| Market: | BioPharma |
| Keywords: | Acclaim PolarAdvantage II (PA2), beta-carotene, chlorophyll, Chlorophyll a, Chlorophyll b, HPLC, Spinach leaf, UltiMate 3000 RSLC, UV detection |
| Matrix: | Acetone extract of spinach leaf |
For Research Use Only. Not for use in diagnostic procedures.