Find methods for your needs
High resolution triglyceride profile of cooking oil using a Thermo Scientific Acclaim C30 column vs a Thermo Scientific Acclaim 120 C18 column with charged aerosol detection
Description
The Thermo Scientific Acclaim C30 5 µm column provides higher resolution than the Thermo Scientific Acclaim 120 C18 5 µm column for the triglyceride profile from cooking oil. The detailed fingerprint was obtained with a 70 minutes run time with detection by the Thermo Scientific Dionex Corona Ultra charged aerosol detector.| Market: | Food and Beverage |
| Keywords: | Acclaim 120 C18, Acclaim C30, Charged aerosol detector, Cooking oil, Peanut oil, Triglycerides |
| Matrix: | Peanut oil |
For Research Use Only. Not for use in diagnostic procedures.