Find methods for your needs
Improved analysis of phospholipids and triglycerides in egg lecithin using a Thermo Scientific Acclaim C30 column with charged aerosol detection
Description
The profile of egg lecithin was determined using a Thermo Scientific Acclaim C30 5 µm column with Thermo Scientific Dionex Corona Ultra charged aerosol detection. It indicates that lecithin from egg yolk contains both phospholipids and triglycerides with other minor impurities.| Market: | BioPharma |
| Keywords: | Acclaim C30, Charged aerosol detector, Egg lecithin, Phospholipids, Triglycerides |
| Matrix: | Egg lecithin |
For Research Use Only. Not for use in diagnostic procedures.