Find methods for your needs
Rapid determination of the components of artificial vanilla flavor on a Thermo Scientific Acclaim PolarAdvantage II (PA2) column
Description
The Thermo Scientific Acclaim PA2 3 µm column is used to separate the components of artificial vanilla by HPLC-UV. With a total run time of less than 3.5 minutes the method allows fast detection of the components present.| Market: | Food and Beverage |
| Keywords: | Acclaim PA2, Artificial vanilla flavor, Benzoic acid, Coumarin, Ethyl vanillin, HPLC, Vanillin |
| Matrix: | Artificial vanilla flavor |
For Research Use Only. Not for use in diagnostic procedures.