Find methods for your needs
Improved analysis of monocarboxylic acids using a Thermo Scientific Acclaim Mixed-Mode WAX-1 column
Description
The Thermo Scientific HPLC system is applied for the analysis of monocarboxylic acids. The separation is performed using a Thermo Scientific Acclaim Mixed-Mode WAX-1 column, with UV detection at 210nm.| Market: | Pharma |
| Keywords: | Acclaim Mixed-Mode WAX-1, Acetic acid, Ascorbic acid, Formic acid, Glycolic acid, HPLC, Iso-ascorbic acid, Lactic acid, Monocarboxylic acids, Propionic acid, Quinic acid, Shikimic acid, UV detection, Vitamin C |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.