Find methods for your needs
Improved analysis of active pharmaceutical ingredients in a pain-relief medicine using a Thermo Scientific Acclaim Mixed-Mode WAX-1 HPLC column Be the first to rate this application
improved analysis active pharmaceutical ingredients a painrelief medicine using a thermo scientific acclaim mixedmode wax1 hplc column
# Compound Name Compound Class RT (min)
1 Caffeine Stimulant 1.37
2 Acetaminophen Analgesic 1.83
3 Salicylamide Analgesic 2.75
4 Acetylsalicyclic acid Analgesic 4.82
5 Salicyclic acid Metabolite 12.41
Description
The Thermo Scientific HPLC system was applied for the analysis of active pharmaceutical ingredients (APIs) in a pain-relief medicine. The separation was performed on a Thermo Scientific Acclaim Mixed-Mode WAX-1 column with UV detection at 220 nm.
Market: Pharma
Keywords: Acetaminophen, Acetyl salicyclic acid, Active pharmaceutical ingredients, API, Aspirin, Caffeine, HPLC, Pain-relief medicine, Paracetamol, Salicylamide, Salycylic acid, UV detection
Matrix: Tablet
Uploaded on 7/14/2014.
For Research Use Only. Not for use in diagnostic procedures.