Find methods for your needs
Improved analysis of tetracyclines in pork using a Thermo Scientific Acclaim PolarAdvantage (PA) HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of tetracyclines in pork. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage (PA) HPLC column, with UV detection at 360 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim Polar Advantage (PA), Antibiotics, Chlortetracycline, Doxycycline, HPLC, Oxytetracycline, Pork, Tetracycline, UV detection |
| Matrix: | Pork extract |
For Research Use Only. Not for use in diagnostic procedures.