Description
In this work, the Thermo Scientific Acclaim Surfactant Plus HPLC column was used to analyse Zonyl FSO fluorosurfactant, a sparingly water-soluble, ethoxylated non-ionic fluorosurfactant. The Acclaim Surfactant Plus column separated the sample based on hydrophobe and degree of ethoxylation simultaneously. Zonyl is a registered trademark of E. I. du Pont de Nemours and Company| Market: | Chemical |
| Keywords: | HPLC, Acclaim Surfactant Plus, Charged aerosol detection, Zonyl FSO Fluorosurfactant |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.