Find methods for your needs
An improved separation of vitamin K isomers using the Thermo Scientific Acclaim C30 HPLC column
Description
In this work the Thermo Scientific Acclaim C30 column provides good resolution between both vitamin K1 and vitamin K2 and corresponding isomers, while the C18 column fails to separate these isomers.| Market: | Pharma |
| Keywords: | Acclaim C30, HPLC, Isomers, UV detection, Vitamin K |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.