Find methods for your needs
Rapid determination of Minoxidil using the Thermo Scientific Acclaim Mixed-Mode WCX-1 HPLC column
Description
The Thermo Scientific Acclaim Mixed-Mode WCX-1 column is used for the fast analysis of minoxidil in formulation with a run time of less than 3 minutes.| Market: | Pharma |
| Keywords: | Acclaim Mixed-Mode WCX-1, HPLC, Minoxidil, UV detection |
| Matrix: | Formulation |
For Research Use Only. Not for use in diagnostic procedures.