Find methods for your needs
Analysis of omeprazole in an anti-heartburn tablet according to the USP monograph using a Thermo Scientific Acclaim PolarAdvantage II HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of omeprazole according to the USP monograph. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage II HPLC column, with UV detection at 305 nm. Since the USP method operates at pH 9, a column with good alkali resistance is needed. The Thermo Scientific Acclaim PolarAdvantage II gives excellent performance for this assay.
Market: Pharma
Keywords: Acclaim PolarAdvantage II (PA2), HPLC, Omeprazole, USP monograph, UV detection
Matrix: Tablet
Uploaded on 5/28/2014.
For Research Use Only. Not for use in diagnostic procedures.