Description
The Thermo Scientific HPLC system is applied for the analysis of cationic surfactants. The separation is performed on a Thermo Scientific Acclaim Surfactant Plus, with charged aerosol detection. The Acclaim Surfactant Plus column is designed specifically for surfactant analysis. It provides ideal selectivity for separating different surfactant types with a single injection, low column bleed, excellent stability, and shorter analysis time. In particular, it is designed to give excellent peak shapes for cationic surfactants.| Market: | Chemical |
| Keywords: | HPLC, Acclaim Surfactant Plus, Charged aerosol detection, Cationic surfactant |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.