Find methods for your needs
Rapid and sensitive determination of clenbuterol, ractopamine and phenolethanolamine in meat using a Thermo Scientific Acclaim PolarAdvantage HPLC column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the determination of clenbuterol, ractopamine and phenolethanolamine in meat. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage HPLC column after automated on-line solid phase extraction using a Thermo Scientific SolEX HRP cartridge, with UV detection at 210 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim PolarAdvantage (PA), Clenbuterol, HPLC, Meat, Online SPE, Phenolethanolamine, Pork, Ractopamine, SolEx HRP, Solid phase extraction |
| Matrix: | Meat |
For Research Use Only. Not for use in diagnostic procedures.