Find methods for your needs
Rapid analysis of Niflumic Acid using Thermo Scientific SOLA WAX SPE cartridges and a Thermo Scientific Accucore RP-MS HPLC column Be the first to rate this application
Description
The Thermo Scientific Dionex UltiMate 3000 HPLC system is applied for the analysis of niflumic acid in human plasma. The samples were prepared using SOLA WAX solid phase extraction (SPE) cartridges, and the separation performed on a Thermo Scientific Accucore RP-MS HPLC column, with MS/MS detection. For Research Use Only. Not for use in diagnostic procedures.
Market: Pharma
Keywords: LC-MS/MS, MS detection, niflumic acid, SOLA WAX, SPE
Matrix: Human plasma
Uploaded on 6/18/2013.
For Research Use Only. Not for use in diagnostic procedures.