Find methods for your needs
Fast analysis of 3 nonsteroidal anti-inflammatory drugs (NSAIDs) using a Thermo Scientific Accucore Polar Premium HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of nonsteroidal anti-inflammatory drugs (NSAIDs). The separation is performed on a Thermo Scientific Accucore Polar Premium HPLC column, with UV detection at 265 nm.| Market: | Pharma |
| Keywords: | Accucore Polar Premium, HPLC, Ibuprofen, Ketoprofen, Naproxen, NSAID, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.