 Rate
                        Rate
                     Ask the Expert
                        Ask the Expert
                     Suggest an Application
                            Suggest an Application
                         Add to Favorites
                        Add to Favorites
                     Create
                            CreateParts List
 Tell a
                        Tell a Friend
 Print
                        PrintFind methods for your needs
Reaction monitoring of fexofenadine API using a PFP HPLC column
Description
The Thermo Scientific Dionex Ultimate 3000 HPLC system was applied for the reaction monitoring of fexofenadine and its impurities during the synthesis of fexofenadine API. The separation was performed on a Thermo Scientific Accucore PFP HPLC column resolution of 2.5 between fexofenadine and its meta isomer was achieved.| Market: | Pharma | 
| Keywords: | Accucore PFP, Fexofenadine, HPLC, UV detection | 
| Matrix: | Reaction mixture | 
| Author: | Anirudha Rakhe, Prakash Chander | 
| Affiliation: | Thermo Fisher Scientific | 
For Research Use Only. Not for use in diagnostic procedures.