Find methods for your needs
Improved HILIC determination of vitamin C using a Thermo Scientific Hypersil GOLD AX HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of vitamin C. The separation is performed on a Thermo Scientific Hypersil GOLD AX HPLC column in HILIC mode, with UV detection at 240 nm.| Market: | Food and Beverage |
| Keywords: | HILIC, HPLC, Hypersil GOLD AX, UV detection, Vitamin C |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.