Find methods for your needs
Rapid determination of Gestodene in human plasma by SLE-LC-MS/MS using a Thermo Scientific HyperSep SLE and a Thermo Scientific Accucore C18 HPLC column
Description
A simple, rapid and sensitive procedure for the determination of gestodene in human plasma by LC-MS/MS was developed. The drug was isolated from plasma using Thermo Scientific HyperSep SLE and the extracts were separated on a Thermo Scientific Accucore C18 HPLC column under reversed-phase, gradient conditions. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | Accucore C18, Gestodene, Hormones, HyperSep SLE, LC-MS/MS, MS detection, Plasma |
| Matrix: | Human plasma |
For Research Use Only. Not for use in diagnostic procedures.