Find methods for your needs
Demonstration of improved resolution and efficiency for the analysis of fat soluble vitamins using a solid core C18 4 μm HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 RSLC system is applied for the determination of fat soluble vitamins. The separation is performed on a Thermo Scientific Accucore XL C18 HPLC column, with UV detection at 280 nm. Compared to a conventional 5 μm fully porous column using the same experimental conditions, the Accucore XL C18 4 μm HPLC column provides improved efficiency and resolution.| Market: | Food and Beverage |
| Keywords: | Accucore XL C18, Fat soluble vitamins, HPLC, Vitamin D2, Vitamin D3, Vitamin E, Vitamin E acetate, Vitamin K1, Vitamin K2 |
| Matrix: | N/A |
| Author: | Derek Hillbeck, Ken Meadows |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.