Find methods for your needs
Improved analysis of xanthines using a Thermo Scientific Hypersil GOLD aQ HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of xanthines. The separation is performed on a Thermo Scientific Hypersil GOLD aQ HPLC column, with UV detection at 254 nm.| Market: | Chemical |
| Keywords: | Caffeine, HPLC, Hypersil GOLD aQ, Hypoxanthine, Theobromine, Theophylline, UV detection, Xanthine |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.