Find methods for your needs
Fast analysis of Penicillins using a Thermo Scientific Hypersil GOLD CN HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of penicillins. The separation is performed on a Thermo Scientific Hypersil GOLD CN HPLC column, with UV detection at 220 nm.| Market: | Pharma |
| Keywords: | Ampicillin, Antibiotics, HPLC, Hypersil GOLD CN, Impurities, N-acetyl penicillamine, Penicllin G, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.