Find methods for your needs
Fast high temperature analysis of herbicides and metabolites using a Thermo Scientific Hypercarb HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of herbicides and metabolites. The separation is performed on a Thermo Scientific Hypercarb high temperature HPLC column, with UV detection at 215 nm.| Market: | Environmental |
| Keywords: | Ametryn, Atrazine, Herbicides, High temperature, HPLC, Hypercarb, Prometryn, Propanil, Propazine, Simazine, Symetryn, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.