Find methods for your needs
Fast determination of procainamide from human plasma using Thermo Scientific SOLA CX SPE and a Thermo Scientific Syncronis C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of procainamide. The samples are prepared using Thermo Scientific SOLA CX solid phase extraction (SPE) cartridges, and the separation performed on a Thermo Scientific Syncronis C18 HPLC column. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | HPLC, Human Plasma, Procainamide, SOLA CX, SPE, Sulfanilamide, Syncronis C18, UV detection |
| Matrix: | Human plasma |
For Research Use Only. Not for use in diagnostic procedures.