Find methods for your needs
Improved analysis of acids, bases and neutrals using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of acids, bases and neutrals. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 270 nm.| Market: | Chemical |
| Keywords: | 3-Nitrobenzoic acid, Acetylprocainamide, Acids, Bases, Benzyl alcohol, Benzylamine, Chlorocinnamic acid, Dihydroxynaphthalene, HPLC, Hypersil GOLD, Neutrals, Phenol, Pyridine, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.