Find methods for your needs
Fast separation of fat soluble vitamins using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of fat soluble vitamins. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column in less than 8 minutes, with UV detection at 280 nm.Market: | Pharma |
Keywords: | Fat soluble vitamins, HPLC, Hypersil GOLD, UV detection, Vitamin A, Vitamin D2, Vitamin D3, Vitamin E, Vitamin E acetate, Vitamin K1 |
Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.