Find methods for your needs
Rapid determination of Steroids from urine by LC-MS/MS using Thermo Scientific SOLA HRP SPE and a Thermo Scientific Accucore RP-MS HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of steroids from urine. The sample was prepared using SOLA HRP solid phase extraction (SPE), and the separation performed on a Thermo Scientific Accucore RP-MS HPLC column in less than 6 minutes, with MS/MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | 11-α hydroxyprogesterone, Accucore RP-MS, Betamethasone, Corticosterone, Cortisone, Hydrocortisone, LC-MS/MS, MS detection, SOLA HRP, SPE, Steroids, Urine |
| Matrix: | Urine |
For Research Use Only. Not for use in diagnostic procedures.