Find methods for your needs
AB72363: Detection of Common Organic Acids using a Compact Ion Chromatography System Coupled with Mass Spectrometry
Description
This application brief demonstrates a method for detecting common organic acids using a Thermo Scientific™ Dionex™ Integrion™ HPIC™ system with a Thermo Scientific™ Dionex™ IonPac™ AS11-HC-4μm column set and coupled to a Thermo Scientific™ ISQ™ EC single quadrupole mass spectrometer.| Market: | Food and Beverage |
| Keywords: | Animal feed, single quad MS, IC-MS, Integrion HPIC, Dionex IonPac AS11-HC-4μm column, ISQ™ EC Single Quadrupole Mass Spectrometer |
| Matrix: | Animal feed |
| Author: | Beibei Huang; Jingli Hu; Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
| Status: | Proof of Concept |
For Research Use Only. Not for use in diagnostic procedures.