Find methods for your needs
Rapid and sensitive UHPLC screening of food dyes in carbonated beverages using UV/Vis wavelength switching
Description
The analysis of food dyes in carbonated beverages is important as many of these dyes are either controlled substances or are being phased out in certain countries due to their reported adverse health effects. Being able to identify and quantify food dyes in beverages quickly and with high sensitivity is therefore important. Reversed-phase chromatography is an excellent technique for the analysis of dyes. Many dyes are readily soluble in reversed-phase eluents and have strong visible and UV absorbance properties. This method demonstrates the separation of ten contentious dyes.| Market: | Food and Beverage |
| Keywords: | Dyes, Food and Beverage, Sunset yellow FCF, Tartrazine, Food Safety, Vanquish Flex UHPLC System, Hypersil GOLD VANQUISH, beverage analysis |
| Matrix: | Beverage |
| Author: | Aaron Lamb |
| Affiliation: | Thermo Fisher Scientific, Runcorn, UK |
For Research Use Only. Not for use in diagnostic procedures.