Find methods for your needs
Rapid analysis of natural sweeteners found in food and beverages using an advanced UHPLC system
Description
Over the last decade there has been a growing interest in low-calorie alternatives to carbohydrate-based sweeteners. Recent publications have shown a dramatic increase in attention toward natural extracts such as the Stevia rebaudiana plant, not only for its sweetening effect but also for additional health benefits attributed to the plant. The major sweetening components are stevioside, rebaudioside A, rebaudioside C, and dulcoside A, each of which is over 300 times sweeter than sucrose based sweeteners. Because of this they are widely used in beverages and foodstuffs.| Market: | Food and Beverage |
| Keywords: | Food and Beverage, Glycosides, Soft drinks, Sweeteners, Syncronis HILIC, UHPLC, stevioside |
| Matrix: | Beverage |
| Author: | Derek Hillbeck, |
| Affiliation: | Thermo Fisher Scientific, Runcorn, UK |
For Research Use Only. Not for use in diagnostic procedures.