Find methods for your needs
Pesticide residues screening analysis in tea and honey using a Q Exactive Focus High-Resolution Mass Spectrometer
Description
We present a multi-residue instrumental method that can be applied for high-throughput screening and semi-quantitation of pesticide residues in food matrices at or below the current legislative requirements. A high-resolution, accurate-mass mass spectrometer operated in Full Scan – Variable Data-Independent Analysis (FS-vDIA)* mode provided an option for full spectrum filtering, retrospective analysis, and multi-parameter-based compound identification. The method was validated for 328 target pesticides, with an option for the future extension to a larger number. (*vDIA not available in US.)| Market: | Food and Beverage |
| Keywords: | QuEChERS, vDIA, Pesticide residues, Q Exactive Focus, Vanquish Flex |
| Matrix: | Honey, Tea |
| Author: | Laszlo Hollosi, Maciej Bromirski, Michal Godula |
| Affiliation: | Thermo FIsher Scientific |
For Research Use Only. Not for use in diagnostic procedures.