Find methods for your needs
AN363: Using Accelerated Solvent Extraction in Alternative Fuel Research
Description
The Accelerated Solvent Extraction (ASE) provides a fast and efficient way to extract various biomass samples for sugar analysis to determine sample viability for alcohol production. This application note describes methods for the extraction of sugars from biomass samples using acidified solvents (in-cell hydrolysis) and extracting pre-hydrolyzed samples.| Market: | Energy |
| Keywords: | biofuel, ASE, Dionium extraction cell, biomass, ASE 200, ASE 350, ASE Prep DE, Soxhlet Extraction, sugar analysis, in-cell hydrolysis |
| Matrix: | Corn stover, Switchgrass |
For Research Use Only. Not for use in diagnostic procedures.