Find methods for your needs
High temperature analysis of glycerin and glycerides in biodiesel (ASTM D6584) using a Thermo Scientific TraceGOLD TG-5MT GC Column
Description
The Thermo Scientific Trace GC system is applied for the ASTM D6584 determination of glycerol and glycerides in biodiesel. The separation is performed on a Thermo Scientific TraceGOLD TG-5MT 15m x 0.25mm (0.1µm film), with flame ionisation detection.| Market: | Chemical |
| Keywords: | ASTM D6584, Diglycerides, Glycerol, monoglycerides, TG-5MT, Triglycerides |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.