Find methods for your needs
Improved analysis of orange soft drink additives using a Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of orange soft drink additives. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column in less than 8 minutes, with UV detection at 254 nm. Orange juice is a complex matrix and as limited sample preparation has been performed in this analysis some matrix peaks are being detected.| Market: | Food and Beverage |
| Keywords: | Aspartame, Benzoic acid, HPLC, Hypersil GOLD, Orange, Saccharin, Soft drink additivies, UV detection |
| Matrix: | Orange soft drink |
For Research Use Only. Not for use in diagnostic procedures.