Find methods for your needs
Separation of related impurities of Furosemide injection as per BP Monograph.
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the separation of related impurities of Furosemide Injection as per BP Monograph 2007. The separation was performed on a Thermo Scientific Syncronis C8 HPLC column with UV detection at 238 nm. With a total run time of 36 minutes the method allows separation of all related impurities of Furosemide. The obtained Relative retention time of Impurity C, B, A, E and D with respect to Furosemide peak are as 0.39, 0.43, 0.76, 1.28 and 1.5 minutes respectively| Market: | Pharma |
| Keywords: | HPLC, Syncronis C8, UV detection, Furosemide Injection, British pharmacopeia 2007 |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.