Find methods for your needs
In-depth protein glycosylation comparison of a biosimilar candidate by hydrophilic interaction UHPLC
Description
The Thermo Scientific Vanquish H system with fluorescence detection is applied for the glycan profile comparison of a biosimilar candidate produced ‘in house’ using Chinese hamster ovary (CHO) cell line using HILIC-based chromatographic separation on Accucore 150-Amide-HILIC column. Developed methodology offers an excellent analytical platform for glycan profile comparison of multiples lots of the biosimilar product.| Market: | BioPharma |
| Keywords: | Biotherapeutics, Glycosylation, IgG, N-glycans, Monoclonal Antibodies, Trastuzumab, Innovator, Biosimilar, NIBRT, Bioprocessing Research |
| Matrix: | Biosimilar product |
| Author: | Silvia Millán, Anne Trappe, Amy Farrell and Jonathan Bones |
| Affiliation: | NIBRT, Dublin, Ireland |
| Status: | Proof of Concept |
For Research Use Only. Not for use in diagnostic procedures.