Find methods for your needs
AN661: Fast routine analysis of polar pesticides in foods by suppressed ion chromatography and mass spectrometry
Description
We present a reliable and cost-effective IC-MS/MS multi-residue method for high-throughput screening and quantitation of polar pesticide residues and their metabolites in food matrices below the current legislative requirements. In contrast to methods described in the literature, sample preparation is simplified and the use of ion chromatography speeds up the separation.| Market: | Food and Beverage |
| Keywords: | AMPA, Glyphosate, IC-MS, Integrion, TSQ Endura, Pesticide residues, polar pesticides, AN64868 |
| Matrix: | Lettuce, Oranges, Wheat flour |
| Author: | Katerina Boušová, Cees Bruggink, Michal Godula |
| Affiliation: | Thermo FIsher Scientific |