Find methods for your needs
Separation of related impurities from Combination drug of Ezetimibe and Simvastatin
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the separation of related impurities separation of combination drug of Ezetimibe and simvastatin. The separation was performed on a Thermo Scientific Hypersil Gold PFP column with UV detection at 238 nm. With a total run time of 120 minutes the method allows separation of all related impurities of Ezetimibe and Simvastatin.| Market: | Pharma |
| Keywords: | Ezetimibe, HPLC, Simvastatin, Hypersil GOLD PFP, UV detection |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.