Find methods for your needs
Quantitative Analysis of Methylmalonic Acid in Plasma Using a Q Exactive Focus High-Resolution Mass Spectrometer for Clinical Research
Description
Clinical research laboratories perform analysis of methylmalonic acid (MMA) using LC-MS because of that technique’s sensitivity and selectivity. Usually, the quantitative method is developed on a triple quadrupole mass spectrometer. Here we evaluated a method implemented on a Q Exactive Focus ultra-high resolution quadrupole-Orbitrap mass spectrometer for the best selectivity.| Market: | Clinical Research |
| Keywords: | Clinical Research, Methylmalonic acid (MMA), Q Exactive Focus |
| Matrix: | Plasma |
| Author: | Mindy Gao, Joe Di Bussolo, Marta Kozak |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.