Find methods for your needs
Bulk Substance Analysis Using DART Source Coupled to LTQ XL Ion Trap and Q Exactive Focus Mass Spectrometers
Description
Forensic laboratories perform bulk substance identification to support investigations in cases involving possession, trafficking, and manufacturing of illicit drugs. A variety of analytical techniques can be used, but mass spectrometry provides methodologies that produce rapid and confident results. We evaluated two analytical workflows using a DART (Direct Analysis in Real Time) ion source coupled to a LTQ XL ion trap mass spectrometer and a Q Exactive Focus hybrid quadrupole- Orbitrap MS. Pharmaceutical pills were analyzed to demonstrate method performance. For forensic use only.| Market: | Forensics; Toxicology |
| Keywords: | Forensic, DART, Orbitrap, bulk substance, Ion Trap |
| Matrix: | Bulk substances |
| Author: | Marta Kozak, Mari Prieto Conaway, Kristine Van Natta |
| Affiliation: | Thermo Fisher Scientific |