Find methods for your needs
Simultaneous, Fast Analysis of Melamine and Analogues in Pharmaceutical Components Using Q Exactive - Benchtop Orbitrap LC-MS/MS
Description
We demonstrated a simple workflow for the simultaneous analysis of melamine and analogues in pharmaceutical components using a Q Exactive benchtop Orbitrap mass spectrometer. The sensitive, simple and robust method employed the Thermo Fisher Scientific Q Exactive high resolution Orbitrap mass spectrometry coupled with UHPLC and HILIC mode chromatographic separation. Simultaneous detection and quantification of melamine and its analogues in Albumin and Guar gum were achieved with a high degree of confidence.| Market: | Food and Beverage |
| Keywords: | HRAM, FDA LIB 4421, HILIC Column, Q Exactive Orbitrap MS, Ultimate UHPLC |
| Matrix: | Albumin, Guar gum |
| Author: | Kate Comstock; Tim Stratton; Hongxia (Jessica) Wang; Yingying Huang |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.