Find methods for your needs
LC-MS/MS Analysis of EtG and EtS in Dilute Urine on the TSQ Endura Triple Quadrupole MS Be the first to rate this application
lcmsms analysis etg ets dilute urine on tsq endura triple quadrupole ms
Description
We developed a high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) method for the forensic toxicological analysis of EtG and EtS in urine with limits of quantitation (LOQs) of 100 and 50 ng/mL, respectively, using only urine dilution as sample preparation. For forensic use only.
Market: Forensics; Toxicology
Keywords: Ethyl Sulfate, EtG, TSQ Endura, EtS, Ethyl glucuronide, Ion pairing, DHAA (dihexylammonium acetate)
Matrix: Urine
Author: Kristine Van Natta, Marta Kozak
Affiliation: Thermo Fisher Scientific
Uploaded on 9/29/2016.