 Rate
                        Rate
                     Ask the Expert
                        Ask the Expert
                     Suggest an Application
                            Suggest an Application
                         Add to Favorites
                        Add to Favorites
                     Create
                            CreateParts List
 Tell a
                        Tell a Friend
 Print
                        PrintFind methods for your needs
Separation of Related Impurities from Vercuronium Bromide as per BP monograph.
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of Vercuronium Bromide. The separation was performed on a Thermo Scientific Acclaim Polar Advantage-II HPLC column using the method described in BP monograph. The results obtained for Resolution exceeded the criteria stated in the BP. Resolution between Vercuronium Bromide and Impurity-C was 8.97 ( met BP criteria of NLT 1.2), Resolution between Vercuronium Bromide and Impurity-F was 2.15 (met BP criteria of NLT 1.2) and Resolution between Vercuronium Bromide and Impurity-B was 10.78 (met BP criteria NLT 2.0).| Market: | Pharma | 
| Keywords: | HPLC, UV detection, Acclaim Polar Advantage-II, Vercuronium Bromide | 
| Matrix: | N/A | 
| Author: | Manish Singh | 
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India | 
For Research Use Only. Not for use in diagnostic procedures.