Find methods for your needs
Separation of Aggregates, Dimer and Fragments from Rituximab by using Size Exclusion Chromatography.
Description
The Thermo Scientific Dionex UltiMate 3000 LC Bio-LC system is applied for the Size exclusion analysis of Rituximab. The separation was performed on a Thermo Scientific MabPac SEC-1 column with a UV detection at 220 nm. With a run time of 30 minutes, the method allows easy separation of Aggregates and Dimer of Rituximab| Market: | BioPharma |
| Keywords: | HPLC, Dimer, MAbPac SEC-1, PDA detection, Rituximab, Aggregate |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.